siRNA Details
Gene Symbol: St8sia1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011374.2 | NP_035504.2 | 3 | 1079 | ||
GenBank mRNA | AK042207.1 | - | 3 | 935 | ||
AK046188.1 | - | 3 | 1019 | |||
AK052444.1 | - | 3 | 730 | |||
BC024821.1 | - | 3 | 1079 | 5357151 | ||
EF101558.1 | - | 3 | 191 | |||
L38677.1 | - | 3 | 703 | |||
X84235.1 | - | 3 | 426 |
