siRNA Details
Gene Symbol: Slc20a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011394.3 | NP_035524.2 | 8 | 1958 | ||
GenBank mRNA | AK087507.1 | - | 8 | 1783 | ||
AK157206.1 | - | 8 | 1832 | |||
AK170985.1 | - | 8 | 1924 | |||
AK180271.1 | - | 1 | 180 | |||
AK211870.1 | - | 1 | 352 | |||
BC046510.1 | - | 8 | 1881 | 4035182 | ||
BC094562.1 | - | 8 | 2338 | |||
U62560.1 | - | 1 | 166 |
