siRNA Details
Gene Symbol: Slc3a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009205.2 | NP_033231.2 | 7 | 1289 | ||
GenBank mRNA | AK052623.1 | - | 7 | 1260 | ||
AK132275.1 | - | 7 | 1261 | |||
BC013441.1 | - | 7 | 1231 | 4222972 | ||
D88533.1 | - | 7 | 1249 |
