siRNA Details
Gene Symbol: Smarcc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009211.2 | NP_033237.2 | 21, 22 | 2404 | ||
GenBank mRNA | AK077733.1 | - | 5, 6 | 741 | ||
AK143485.1 | - | 21, 22 | 2392 | |||
AK143513.1 | - | 1, 2 | 154 | |||
AK144122.1 | - | 21, 22 | 2378 | |||
AK144631.1 | - | 21, 22 | 2372 | |||
AK145352.1 | - | 21, 22 | 2371 | |||
BC022173.1 | - | 21, 22 | 2368 | |||
BC052423.1 | - | 21, 22 | 2366 | 6400646 | ||
BC053064.1 | - | 21, 22 | 2360 | |||
BC056375.1 | - | 21, 22 | 2367 | |||
U85614.1 | - | 21, 22 | 2359 |
