siRNA Details
Gene Symbol: Ccdc40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175430.4 | NP_780639.1 | 3 | 1467 | ||
GenBank mRNA | AK046971.1 | - | 3 | 1470 | ||
AK054172.1 | - | 4 | 981 | |||
BC072591.1 | - | 5 | 1232 | |||
BC085159.1 | - | 5 | 1227 | 30533905 |
