siRNA Details
Gene Symbol: Sptlc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011479.3 | NP_035609.1 | 2 | 263 | ||
GenBank mRNA | AK044917.1 | - | 2 | 264 | ||
AK057644.1 | - | 2 | 299 | |||
AK089019.1 | - | 2 | 257 | |||
AK159346.1 | - | 2 | 264 | |||
BC003227.1 | - | 2 | 296 | 3483158 | ||
U27455.1 | - | 2 | 231 | |||
X95642.1 | - | 2 | 234 |
