siRNA Details
Gene Symbol: Sptlc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011479.3 | NP_035609.1 | 10 | 1383 | ||
GenBank mRNA | AK044917.1 | - | 10 | 1384 | ||
AK057644.1 | - | 10 | 1419 | |||
AK080181.1 | - | 4 | 1581 | |||
AK089019.1 | - | 10 | 1377 | |||
AK133728.1 | - | 4 | 444 | |||
AK139681.1 | - | 2 | 86 | |||
AK159346.1 | - | 10 | 1384 | |||
AK172965.1 | - | 3 | 179 | |||
BC003227.1 | - | 10 | 1416 | 3483158 | ||
U27455.1 | - | 10 | 1351 | |||
X95642.1 | - | 10 | 1354 |
