siRNA Details
Gene Symbol: Rnf43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172448.3 | NP_766036.2 | 9 | 1674 | ||
GenBank mRNA | AK028750.1 | - | 8 | 1033 | ||
BC029717.1 | - | 1 | 50 | |||
BC037465.1 | - | 1 | 114 | |||
BC075707.1 | - | 7 | 810 | |||
BC131957.1 | - | 7 | 792 | 40130234 |
