siRNA Details
Gene Symbol: Stat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205313.1 | NP_001192242.1 | 21 | 2111 | ||
NM_001205314.1 | NP_001192243.1 | 21 | 2129 | |||
NM_009283.4 | NP_033309.3 | 21 | 2144 | |||
GenBank mRNA | AK018544.1 | - | 21 | 2095 | ||
AK039458.1 | - | 21 | 2145 | |||
AK046517.1 | - | 21 | 2111 | |||
AK082706.1 | - | 21 | 2126 | |||
AK084855.1 | - | 21 | 2078 | |||
AK157263.1 | - | 21 | 1993 | |||
AK159887.1 | - | 21 | 2128 | |||
BC042551.1 | - | 4 | 276 | |||
U06924.1 | - | 18 | 1798 |
