siRNA Details
Gene Symbol: Fam20a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153782.1 | NP_722477.1 | 10, 11 | 1893 | ||
GenBank mRNA | AK142932.1 | - | 10, 11 | 1918 | ||
BC011274.1 | - | 10, 11 | 1797 | |||
BC023310.1 | - | 3, 4 | 194 | |||
BC029169.1 | - | 10, 11 | 1893 | 4947019 |
