siRNA Details
Gene Symbol: Stk11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB015801.1 | - | 8 | 1096 | ||
AF129870.1 | - | 8 | 1474 | |||
AF145287.1 | - | 8 | 1078 | |||
AF151711.1 | - | 8 | 1078 | |||
AK004499.1 | - | 6 | 757 | |||
AK147802.1 | - | 8 | 1138 | |||
AK171909.1 | - | 9 | 1872 | |||
AK172385.1 | - | 8 | 1861 | |||
AK172528.1 | - | 8 | 1858 | |||
BC052379.1 | - | 8 | 1139 | 5688695 | ||
EU730638.1 | - | 8 | 1057 |
