siRNA Details
Gene Symbol: Dis3l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172157.1 | NP_001165628.1 | 12 | 1434 | ||
NM_153530.2 | NP_705758.1 | 11 | 1392 | |||
GenBank mRNA | AK029652.1 | - | 12 | 1448 | ||
AK031180.1 | - | 12 | 1417 | |||
BC036177.1 | - | 11 | 1353 | 5004341 |
