siRNA Details
Gene Symbol: Adamts18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172466.2 | NP_766054.2 | 20 | 3406 | ||
GenBank mRNA | AK036137.1 | - | 17 | 2485 | ||
AK053851.1 | - | 19 | 3518 | |||
AK140876.1 | - | 20 | 3182 | |||
AK163023.1 | - | 5 | 634 | |||
BC094674.1 | - | 20 | 3061 | 30548647 | ||
BC144816.1 | - | 18 | 2664 | |||
BC144817.1 | - | 17 | 2536 |
