siRNA Details
Gene Symbol: Adamts18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172466.2 | NP_766054.2 | 13 | 2362 | ||
GenBank mRNA | AK036137.1 | - | 10 | 1441 | ||
AK053851.1 | - | 12 | 2474 | |||
AK140876.1 | - | 13 | 2138 | |||
BC094674.1 | - | 13 | 2017 | 30548647 | ||
BC144816.1 | - | 11 | 1620 | |||
BC144817.1 | - | 11 | 1623 |
