siRNA Details
Gene Symbol: Zfp280c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166648.1 | NP_001160120.1 | 6 | 501 | ||
NM_001166649.1 | NP_001160121.1 | 6 | 522 | |||
NM_001166650.1 | NP_001160122.1 | 5 | 460 | |||
NM_153532.3 | NP_705760.2 | 5 | 832 | |||
GenBank mRNA | AK129399.1 | - | 6 | 481 | ||
AK141491.1 | - | 6 | 502 | |||
BC028839.1 | - | 5 | 472 | 5150255 | ||
BC051397.1 | - | 5 | 460 | 5150255 | ||
BC063775.1 | - | 5 | 844 | 5375514 |
