siRNA Details
Gene Symbol: Fam83b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001045518.1 | NP_001038983.1 | 4 | 1580 | ||
GenBank mRNA | AK049628.1 | - | 5 | 1672 | ||
AK145108.1 | - | 5 | 1665 | |||
BC120577.1 | - | 4 | 1580 | 40129500 | ||
BC120579.1 | - | 4 | 1580 | 40129502 |
