siRNA Details
Gene Symbol: Fam83b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001045518.1 | NP_001038983.1 | 2, 3 | 639 | ||
GenBank mRNA | AK049628.1 | - | 3, 4 | 731 | ||
AK145108.1 | - | 3, 4 | 723 | |||
BC120577.1 | - | 2, 3 | 639 | 40129500 | ||
BC120579.1 | - | 2, 3 | 639 | 40129502 |
