siRNA Details
Gene Symbol: Tmem87a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110496.1 | NP_001103966.1 | 16 | 1530 | ||
NM_001110497.1 | NP_001103967.1 | 16 | 1530 | |||
NM_173734.3 | NP_776095.2 | 16 | 1533 | |||
GenBank mRNA | AK044591.1 | - | 16 | 1484 | ||
AK134268.1 | - | 16 | 1496 | |||
BC004047.1 | - | 16 | 1479 | |||
BC024676.1 | - | 6 | 481 | |||
BC027354.1 | - | 16 | 1506 | 4010477 |
