siRNA Details
Gene Symbol: Armc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034858.3 | NP_001030030.2 | 8, 9 | 1233 | ||
GenBank mRNA | AK131686.1 | - | 4, 5 | 428 | ||
AK141039.1 | - | 2, 3 | 1565 | |||
BC172168.1 | - | 3, 4 | 975 |
