siRNA Details
Gene Symbol: Plekhh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177606.4 | NP_808274.2 | 16 | 2567 | ||
GenBank mRNA | AK029252.1 | - | 16 | 2569 | ||
AK173337.1 | - | 13 | 2222 | |||
BC125583.1 | - | 16 | 2532 | 40130098 | ||
BC137804.1 | - | 16 | 2529 | 8860823 |
