siRNA Details
Gene Symbol: Tbx3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011535.3 | NP_035665.2 | 1 | 798 | ||
NM_198052.2 | NP_932169.1 | 2 | 994 | |||
GenBank mRNA | AK031708.1 | - | 1 | 1315 | ||
AK034758.1 | - | 1 | 1358 | |||
AK052453.1 | - | 1 | 1013 | |||
AK145436.1 | - | 1 | 715 | |||
AK157247.1 | - | 1 | 559 | |||
AK172118.1 | - | 1 | 559 | |||
BC096551.1 | - | 2 | 994 | 30547736 |
