siRNA Details
Gene Symbol: Tbx3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011535.3 | NP_035665.2 | 1 | 831 | ||
NM_198052.2 | NP_932169.1 | 2 | 1027 | |||
GenBank mRNA | AK031708.1 | - | 1 | 1348 | ||
AK034758.1 | - | 1 | 1391 | |||
AK052453.1 | - | 1 | 1046 | |||
AK145436.1 | - | 1 | 748 | |||
AK157247.1 | - | 1 | 592 | |||
AK172118.1 | - | 1 | 592 | |||
BC096551.1 | - | 2 | 1027 | 30547736 |
