siRNA Details
Gene Symbol: Tbxas1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011539.3 | NP_035669.3 | 9 | 1123 | ||
GenBank mRNA | AK048544.1 | - | 9 | 1105 | ||
AK085414.1 | - | 6 | 875 | |||
AK139479.1 | - | 9 | 1086 | |||
BC029014.1 | - | 9 | 1125 | 5007629 | ||
L18868.1 | - | 9 | 994 |
