siRNA Details
Gene Symbol: Ago3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153402.2 | NP_700451.2 | 18 | 2690 | ||
GenBank mRNA | AB081473.1 | - | 18 | 2542 | ||
AK035042.1 | - | 18 | 2654 | |||
AK083652.1 | - | 18 | 2658 | |||
AK154436.1 | - | 18 | 2691 | |||
AK171118.1 | - | 18 | 2691 | |||
BC137964.1 | - | 18 | 2537 | 8860983 | ||
BC145061.1 | - | 18 | 2514 | |||
BC145063.1 | - | 17 | 2400 |
