siRNA Details
Gene Symbol: Zeb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011546.3 | NP_035676.1 | 7 | 2682 | ||
GenBank mRNA | BC139768.1 | - | 7 | 2651 | 40111224 | |
BC139769.1 | - | 7 | 2651 | 40111227 | ||
D76432.1 | - | 7 | 2651 | |||
U26259.1 | - | 7 | 2668 |
