siRNA Details
Gene Symbol: Tfap2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025305.2 | NP_001020476.1 | 6 | 1293 | ||
NM_001286340.1 | NP_001273269.1 | 6 | 1258 | |||
NM_009334.3 | NP_033360.2 | 6 | 1258 | |||
GenBank mRNA | AK005128.1 | - | 6 | 1214 | ||
AK017373.1 | - | 6 | 1260 | |||
AK028906.1 | - | 6 | 1213 | |||
AK032045.1 | - | 6 | 1295 | |||
AK078046.1 | - | 5 | 947 | |||
X78197.1 | - | 6 | 1184 |
