siRNA Details
Gene Symbol: A430105I19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001982.2 | NP_001001982.2 | 5 | 634 | ||
GenBank mRNA | AK040533.1 | - | 4 | 675 | ||
AK137584.1 | - | 5 | 636 | |||
BC075684.1 | - | 4 | 386 | 30611649 |
