siRNA Details
Gene Symbol: Adamtsl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029967.3 | NP_084243.3 | 25 | 4875 | ||
GenBank mRNA | AK031246.1 | - | 8 | 1478 | ||
AK132420.1 | - | 13 | 3315 | |||
BC116295.1 | - | 8 | 1469 | |||
BC116296.1 | - | 8 | 1469 |
