siRNA Details
Gene Symbol: Rap1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024457.2 | NP_077777.1 | 6 | 585 | ||
GenBank mRNA | AK143910.1 | - | 6 | 568 | ||
AK153870.1 | - | 6 | 542 | |||
AK163242.1 | - | 6 | 649 | |||
AK167548.1 | - | 6 | 541 | |||
AK205627.1 | - | 2 | 114 | |||
BC033382.1 | - | 6 | 525 | |||
BC052480.1 | - | 6 | 585 | |||
BC094066.1 | - | 6 | 525 | |||
BC147390.1 | - | 6 | 457 | 9056464 | ||
BC147391.1 | - | 6 | 457 | 9056441 | ||
BC147700.1 | - | 5 | 443 | 9087459 | ||
BC147713.1 | - | 5 | 443 | 9087472 | ||
CK793274.1 | - | 1 | 102 |
