siRNA Details
Gene Symbol: Rap1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024457.2 | NP_077777.1 | 2 | 181 | ||
GenBank mRNA | AK143910.1 | - | 2 | 164 | ||
AK153870.1 | - | 2 | 138 | |||
AK163242.1 | - | 2 | 245 | |||
AK167548.1 | - | 2 | 137 | |||
AK178034.1 | - | 2 | 54 | |||
AK196093.1 | - | 2 | 54 | |||
AK199293.1 | - | 2 | 54 | |||
AK218552.1 | - | 2 | 54 | |||
BC033382.1 | - | 2 | 121 | |||
BC052480.1 | - | 2 | 181 | |||
BC094066.1 | - | 2 | 121 | |||
BC147390.1 | - | 2 | 53 | 9056464 | ||
BC147391.1 | - | 2 | 53 | 9056441 | ||
BC147700.1 | - | 1 | 39 | 9087459 | ||
BC147713.1 | - | 1 | 39 | 9087472 | ||
BU848028.1 | - | 186 |
