siRNA Details
Gene Symbol: Mcu | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033259.4 | NP_001028431.2 | 6 | 850 | ||
GenBank mRNA | AK161322.1 | - | 3 | 672 | ||
BC139254.1 | - | 3 | 467 | 8862276 | ||
BC139257.1 | - | 3 | 467 | 8862279 | ||
DV649183.1 | - | 1 | 121 |
