siRNA Details
Gene Symbol: Gls2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033264.3 | NP_001028436.2 | 17 | 1717 | ||
NM_001285777.1 | NP_001272706.1 | 16 | 1637 | |||
NM_001285779.1 | NP_001272708.1 | 17 | 1746 | |||
GenBank mRNA | AK039618.1 | - | 16 | 1525 | ||
AK165896.1 | - | 17 | 1663 | |||
AK168761.1 | - | 10 | 849 | |||
AK220231.1 | - | 17 | 1669 | |||
BC141115.1 | - | 17 | 1717 | 9055727 | ||
EU770627.1 | - | 17 | 1655 |
