siRNA Details
Gene Symbol: Gls2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033264.3 | NP_001028436.2 | 13, 14 | 1441 | ||
NM_001285777.1 | NP_001272706.1 | 12, 13 | 1361 | |||
NM_001285779.1 | NP_001272708.1 | 13, 14 | 1470 | |||
GenBank mRNA | AK039618.1 | - | 12, 13 | 1249 | ||
AK165896.1 | - | 13, 14 | 1387 | |||
AK168761.1 | - | 6, 7 | 573 | |||
AK220231.1 | - | 13, 14 | 1393 | |||
BC141115.1 | - | 13, 14 | 1441 | 9055727 |
