siRNA Details
Gene Symbol: Wdpcp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145425.3 | NP_663400.2 | 10 | 1156 | ||
GenBank mRNA | AF424701.1 | - | 10 | 1119 | ||
AK043672.1 | - | 9 | 1043 | |||
AK083042.1 | - | 10 | 1157 | |||
BC141166.1 | - | 10 | 1120 | 9055778 |
