siRNA Details
Gene Symbol: Wdpcp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145425.3 | NP_663400.2 | 10 | 1093 | ||
GenBank mRNA | AF424701.1 | - | 10 | 1056 | ||
AK043672.1 | - | 9 | 980 | |||
AK083042.1 | - | 10 | 1094 | |||
BC141166.1 | - | 10 | 1057 | 9055778 |
