siRNA Details
Gene Symbol: Clint1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001045520.3 | NP_001038985.2 | 5 | 597 | ||
GenBank mRNA | AB093212.1 | - | 4 | 1684 | ||
AK147875.1 | - | 5 | 582 | |||
AK168991.1 | - | 5 | 563 | |||
BC004080.1 | - | 2 | 135 |
