siRNA Details
Gene Symbol: Rufy1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172557.2 | NP_766145.1 | 9 | 1105 | ||
GenBank mRNA | AJ250024.1 | - | 8 | 781 | ||
AK046625.1 | - | 9 | 1122 | |||
AK046633.1 | - | 9 | 1106 | |||
AK051178.1 | - | 9 | 1111 |
