siRNA Details
Gene Symbol: Tectb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009348.3 | NP_033374.2 | 5, 6 | 709 | ||
GenBank mRNA | BC138888.1 | - | 5, 6 | 638 | 8861910 | |
BC138889.1 | - | 5, 6 | 638 | 8861911 | ||
X99806.2 | - | 5, 6 | 709 |
