siRNA Details
Gene Symbol: Tef | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017376.3 | NP_059072.1 | 4 | 864 | ||
NM_153484.3 | NP_705617.2 | 4 | 771 | |||
GenBank mRNA | AF194420.1 | - | 4 | 798 | ||
AK042146.1 | - | 4 | 1025 | |||
AK141606.1 | - | 4 | 692 | |||
AK145125.1 | - | 4 | 833 | |||
AK147965.1 | - | 4 | 797 | |||
AK154472.1 | - | 4 | 809 | |||
AY540631.1 | - | 4 | 850 | |||
AY540632.1 | - | 4 | 665 | |||
BC017689.1 | - | 4 | 627 | 4242534 | ||
BC036982.1 | - | 4 | 828 | 4976241 |
