siRNA Details
Gene Symbol: Taok1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144825.2 | NP_659074.2 | 7 | 1213 | ||
GenBank mRNA | AK138443.1 | - | 1185 | |||
AK143530.1 | - | 7 | 1220 | |||
BC047271.1 | - | 6 | 560 | |||
BC151012.1 | - | 7 | 1007 | 9087925 | ||
BC151015.1 | - | 7 | 1007 | 9087928 | ||
BC151016.1 | - | 7 | 1006 | 9087929 | ||
CF164486.1 | - | 3 | 201 | |||
CN717636.1 | - | 6 | 567 |
