siRNA Details
Gene Symbol: Rnf157 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027258.1 | NP_081534.1 | 9 | 996 | ||
GenBank mRNA | AK030994.1 | - | 5 | 235 | ||
AK038042.1 | - | 3 | 223 | |||
AK169566.1 | - | 4 | 176 | |||
AK171736.1 | - | 4 | 218 | |||
BC053070.1 | - | 9 | 775 | |||
CF539215.1 | - | 8 | 638 |
