siRNA Details
Gene Symbol: Tnrc6c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198022.2 | NP_932139.2 | 5 | 865 | ||
GenBank mRNA | AK147471.1 | - | 6 | 863 | ||
AK147551.1 | - | 5 | 581 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tnrc6c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198022.2 | NP_932139.2 | 5 | 865 | ||
GenBank mRNA | AK147471.1 | - | 6 | 863 | ||
AK147551.1 | - | 5 | 581 |