siRNA Details
Gene Symbol: Nol10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008421.1 | NP_001008421.1 | 10 | 763 | ||
GenBank mRNA | AK148210.1 | - | 10 | 779 | ||
AK160864.1 | - | 10 | 799 | |||
AK164400.1 | - | 10 | 789 | |||
AK166160.1 | - | 10 | 792 | |||
BC086676.1 | - | 10 | 763 | 30659737 |
