siRNA Details
Gene Symbol: Ptpdc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301781.1 | NP_001288710.1 | 6, 7 | 930 | ||
NM_207232.2 | NP_997115.1 | 5, 6 | 747 | |||
GenBank mRNA | AK030415.1 | - | 5, 6 | 740 | ||
AK038922.1 | - | 5, 6 | 660 | |||
AK053480.1 | - | 5, 6 | 518 | |||
AK132781.1 | - | 6, 7 | 839 | |||
BC066081.1 | - | 5, 6 | 648 | 30355705 |
