siRNA Details
Gene Symbol: Tdrd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253755.1 | NP_001240684.1 | 4 | 551 | ||
NM_172605.3 | NP_766193.3 | 4 | 551 | |||
GenBank mRNA | AK032892.1 | - | 4 | 496 | ||
AK033390.1 | - | 4 | 496 | |||
BC005670.1 | - | 4 | 452 | 3153274 | ||
BC066144.1 | - | 4 | 499 | 30435963 |
