siRNA Details
Gene Symbol: Crisp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204071.1 | NP_001191000.1 | 5 | 355 | ||
NM_009420.2 | NP_033446.1 | 5 | 373 | |||
GenBank mRNA | AK018856.1 | - | 5 | 304 | ||
BC049615.1 | - | 5 | 325 | 6742033 | ||
CJ046590.1 | - | 5 | 296 | |||
M25533.1 | - | 5 | 325 |
