siRNA Details
Gene Symbol: Traf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009421.3 | NP_033447.2 | 9 | 1252 | ||
GenBank mRNA | AK029135.1 | - | 9 | 1254 | ||
AK145075.1 | - | 827 | ||||
AK169515.1 | - | 9 | 1045 | |||
AK172598.1 | - | 5 | 856 | |||
BC096595.1 | - | 5 | 818 | 30748947 | ||
L35302.1 | - | 8 | 998 |
