siRNA Details
Gene Symbol: Traf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011633.2 | NP_035763.2 | 1999 | |||
GenBank mRNA | AK144694.1 | - | 1950 | |||
BC012702.1 | - | 1953 | 4218625 | |||
BC038645.1 | - | 1145 | ||||
D78141.1 | - | 1976 | ||||
D83528.1 | - | 1841 |

More Information
Other Species Targeted

Gene Ontology Categories:
