siRNA Details
Gene Symbol: Trpc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253682.1 | NP_001240611.1 | 3 | 889 | ||
NM_001253683.1 | NP_001240612.1 | 3 | 889 | |||
NM_016984.3 | NP_058680.1 | 3 | 889 | |||
GenBank mRNA | AF011543.1 | - | 3 | 706 | ||
AF019663.1 | - | 3 | 706 | |||
AF190646.1 | - | 3 | 780 | |||
AK044265.1 | - | 3 | 798 | |||
AK049788.1 | - | 3 | 1023 | |||
AK080670.1 | - | 3 | 890 | |||
AK084139.1 | - | 3 | 882 | |||
BC057335.1 | - | 3 | 780 | |||
BC060160.1 | - | 3 | 780 | |||
BC120729.1 | - | 3 | 804 | 40129652 | ||
BC131915.1 | - | 3 | 804 | 40130192 | ||
U50921.1 | - | 3 | 842 | |||
U50922.1 | - | 3 | 842 |
