siRNA Details
Gene Symbol: Tsc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039363.2 | NP_001034452.1 | 37 | 4960 | ||
NM_001286713.1 | NP_001273642.1 | 39 | 5158 | |||
NM_001286714.1 | NP_001273643.1 | 38 | 5084 | |||
NM_001286716.1 | NP_001273645.1 | 37 | 4955 | |||
NM_001286718.1 | NP_001273647.1 | 37 | 4957 | |||
NM_001286720.1 | NP_001273649.1 | 39 | 5158 | |||
NM_011647.3 | NP_035777.2 | 38 | 5089 | |||
GenBank mRNA | AF132986.1 | - | 35 | 4686 | ||
AK143799.1 | - | 37 | 4959 | |||
AK147302.1 | - | 38 | 5030 | |||
AK147487.1 | - | 39 | 5127 | |||
AK147874.1 | - | 37 | 4924 | |||
AK147908.1 | - | 38 | 5090 | |||
AK148062.1 | - | 38 | 5081 | |||
AK148271.1 | - | 25 | 3529 | |||
AK163991.1 | - | 3 | 308 | |||
AK164586.1 | - | 3 | 238 | |||
AK170589.1 | - | 37 | 4951 | |||
BC052449.1 | - | 37 | 4936 | 6409037 | ||
BC060701.1 | - | 37 | 4936 | 6409037 | ||
U37775.1 | - | 37 | 5001 | |||
U39818.1 | - | 39 | 5129 |
